missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD2AP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA528391
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recommended positive controls: 293T, A431, HeLa, HepG2, A375, Rat2, K562, THP-1, HL-60. Predicted reactivity: Mouse (87%), Rat (86%), Pig (92%), Rhesus Monkey (96%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a scaffolding molecule that regulates the actin cytoskeleton. The protein directly interacts with filamentous actin and a variety of cell membrane proteins through multiple actin binding sites, SH3 domains, and a proline-rich region containing binding sites for SH3 domains. The cytoplasmic protein localizes to membrane ruffles, lipid rafts, and the leading edges of cells. It is implicated in dynamic actin remodeling and membrane trafficking that occurs during receptor endocytosis and cytokinesis. Haploinsufficiency of this gene is implicated in susceptibility to glomerular disease.Especificaciones
CD2AP | |
Polyclonal | |
Unconjugated | |
CD2AP | |
adapter protein CMS; AL024079; C78928; Cas ligand with multiple SH3 domains; Cas ligand with multiple Src homology 3 (SH3) domains; CD2 associated protein; Cd2ap; CD2-associated protein; CMS; mesenchyme to epithelium transition protein, SH3 domains; mesenchyme-to-epithelium transition protein with SH3 domains 1; Mets1; METS-1; SH3 domain-containing adapter protein; SH3 domain-containing adaptor protein METS-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12488, 23607, 316258 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
F1LRS8, Q9JLQ0, Q9Y5K6 | |
CD2AP | |
Recombinant fragment corresponding to a region within amino acids 1 and 281 of Human CD2AP | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |