missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD264 (TRAIL-R4) Monoclonal Antibody (TRAIL-R4-01), PE, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA110164
Detalles adicionales : Peso : 0.01000kg
Descripción
TRAIL (also known as Apo-2L) is a member of the tumor necrosis factor (TNF) ligand family that rapidly induces apoptosis in a variety of transformed cell lines. A fourth member of the emerging TRAIL receptor family, TRAIL-R4, has signaling capacity similar to that of TRAIL-R1 and TRAIL-R2 with respect to NF-kappaB activation, but differs in its inability to induce apoptosis. Like TRAIL-R1 and TRAIL-R2, this receptor shows widespread tissue expression. The human TRAIL-R4 gene has been mapped to chromosome 8p22-21, clustered with three other TRAIL receptors.Especificaciones
CD264 (TRAIL-R4) | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Q9UBN6 | |
TNFRSF10D | |
TRAIL-R4 (aa 1-210) - hIgGhc fusion protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
TRAIL-R4-01 | |
PE | |
TNFRSF10D | |
CD264; DCR2; Decoy receptor 2; TNF receptor superfamily member 10d; TNF receptor-related receptor for TRAIL; TNF-related apoptosis-inducing ligand receptor 4; TNFRSF10D; TRAIL receptor 4; TRAIL receptor with a truncated death domain; TRAILR4; TRAIL-R4; TRUNDD; Tumor necrosis factor receptor superfamily member 10D; tumor necrosis factor receptor superfamily member 10d, decoy with truncated death domain; tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain; UNQ251/PRO288 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
8793 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |