Learn More
CD146 Mouse anti-Human, Alexa Fluor 647, Clone: P1H12, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 563619
Detalles adicionales : Peso : 0.01000kg
Descripción
The P1H12 monoclonal antibody specifically binds to CD146. CD146 is a 118 kDa transmembrane glycoprotein also known as MCAM, MUC18, or Mel-CAM. CD146 is a member of the immunoglobulin superfamily and is expressed by angioblasts and mesenchymal stems cells and is strongly expressed by blood vessel endothelium and smooth muscle. CD146 is also expressed by melanoma cells, intermediate trophoblasts and a subpopulation of activated T cells. The P1H12 monoclonal antibody has been reported to block endothelial cell adhesion that is observed very early in embryogenesis. It can be useful in the study of embryologic vasculogenesis. This antibody is suitable for immunohistochemical staining of acetone-fixed frozen tissue sections, immunoprecipitation and ELISA.
Especificaciones
CD146 | |
Monoclonal | |
Alexa Fluor 647 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Umbilical Vein Cells | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
P1H12 | |
MCAM; MELCAM; MUC18; Gicerin; Melanoma cell adhesion molecule | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |