missing translation for 'onlineSavingsMsg'
Learn More

CD194 Mouse anti-Human, PE-Cy7, Clone: 1G1, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 557864

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15899078

  • 590.00€ / cada uno
Fecha estimada de envío: 15-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The monoclonal antibody 1G1 reacts with the human CC chemokine receptor, CCR4. CCR4 (previously known as K5-5), a seven-transmembrane, G-protein-coupled receptor, is the specific receptor for CC chemokines, MDC/CCL22 and TARC/CC17. It has been shown that CCR4 mRNA is expressed mainly in thymus and spleen. CCR4 mRNA was also detected in peripheral T-lymphocytes, basophils, monocytes and platelets. In addition, CCR4 has been shown to be expressed selectively on polarized Th2 cells. The human CCR4 gene has been mapped to chromosome 3p24. The immunogen used to generate 1G1 hybridoma was human CCR4-transfected L1.2 mouse lymphoma cells. This antibody is NOT a neutralizing antibody.

Host Species: Mouse
Clone: 1G1
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Human CCR4-Transfected L1.2 Mouse Lymphoma

Flow Cytometry, Immunofluorescence

Especificaciones

Especificaciones

CD194
Monoclonal
0.2mg/mL
CCR4; C-C chemokine receptor type 4; CMKBR4; K5-5
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
1G1
PE-Cyanine7
Aqueous buffered solution containing ≤0.09% sodium azide.
Human CCR4 Transfected Cell Line
0.1 mg
Immunology
Human
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.