Learn More
5-Lipoxygenase Mouse, Unlabeled, Clone: 33, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 610695
Detalles adicionales : Peso : 0.01000kg
Descripción
5-Lipoxygenase (5-LO) is the initial enzyme that converts arachidonic acid to leukotrienes, which are important inflammatory and vasoconstrictive metabolites. It is activated in response to a number of stimuli, such as differentiation and allergen challenges. The 5-LO gene, abundantly expressed in placenta, lung, and leukocytes, encodes a protein of 674 amino acids with an apparent molecular weight of 78kDa. 5-LO is a Ca2+ and ATP-dependent enzyme that translocates from the cytosol to either a nuclear or plasma membrane compartment following activation. A proline-rich domain of 5-LO (amino acids 566-577) has been identified as a binding site for the PTyr-binding protein, Grb2. This Grb2 site links tyrosine kinases with activation and redistribution of 5-LO. Furthermore, tyrosine kinase inhibitors increase the activity of 5-LO and block the enzyme's subcellular redistribution.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Especificaciones
5-Lipoxygenase | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
and ≤0.09% sodium azide., glycerol, Aqueous buffered solution containing BSA | |
150 μg | |
Cell Biology | |
Rat, Mouse, Human, Chicken | |
IgG1 |
Western Blot | |
33 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.