missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ANP32A Recombinant Protein
Marca: Abnova™ H00008125-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
AAH07200 | |
Solution | |
8125 | |
ANP32A (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
C15orf1/I1PP2A/LANP/MAPM/MGC119787/MGC150373/PHAP1/PHAPI/PP32 | |
ANP32A | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
53.13 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANP32A | |
Human | |
Yes |