Learn More
Annexin V Antibody (S08-3A4), Novus Biologicals™
Código de producto. 18609974 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18609974 50 μg 50 microlitros
18634125 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18609974 Proveedor Novus Biologicals N.º de proveedor NBP31954350ul
Rabbit Monoclonal Antibody
Annexin V Monoclonal antibody specifically detects Annexin V in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Annexin V
Aplicaciones Western Blot, Immunofluorescence
Clasificación Monoclonal
Clon S08-3A4
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Alias de gen Anchorin CII, annexin A5, Annexin V, annexin-5, ANX5, Calphobindin I, Endonexin II, ENX2CBP-I, Lipocortin V, PAP-I, Placental anticoagulant protein 4, Placental anticoagulant protein I, PP4, Thromboplastin inhibitor, VAC-alpha, Vascular anticoagulant-alpha
Especie del huésped Rabbit
Inmunógeno A synthetic peptide of human Annexin V (Uniprot # P08758)
Método de purificación Affinity purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Apoptosis, Apoptosis Detections
Primario o secundario Primary
ID de gen (Entrez) 308
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG