missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Annexin A1 |
| Aplicaciones | Western Blot, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | ANXA1/3869R |
| Conjugado | Alexa Fluor 594 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant full-length human Annexin A1 protein (Uniprot: P04083) |
| Método de purificación | Protein A or G purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?