missing translation for 'onlineSavingsMsg'
Learn More

Angiopoietin-2 Antibody (5A9), Novus Biologicals™

Código de producto. 30043102 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
100 μL
50 μL
Unit Size:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30043102 100 μL 100 microlitros
30043654 50 μL 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30043102 Supplier Novus Biologicals Supplier No. NBP326508100ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Human Monoclonal Antibody

Angiopoietin-2 Monoclonal antibody specifically detects Angiopoietin-2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Angiopoietin-2
Aplicaciones ELISA
Clasificación Monoclonal
Clon 5A9
Conjugado Unconjugated
Dilución ELISA
Formulación 50% Glycerol, 0.01M PBS, PH 7.4
Alias de gen AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand
Especie del huésped Human
Inmunógeno Recombinant Human Angiopoietin-2 protein [UniProt O15123]
Método de purificación Affinity purified
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Angiogenesis, Cancer
Primario o secundario Primary
ID de gen (Entrez) 285
Especies diana Human
Contenido y almacenamiento Store at -20 to -70C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.