Learn More
Invitrogen™ Human AMPK alpha-1/beta-1/gamma-2, His Tag Recombinant Protein
Descripción
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant full-length human AMPK (combination of A1/B1/G2 subunits) was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag. Validated in LanthaScreen™ Kinase Binding Assay Protocols available for simple use and optimized performance Multiple pack sizes available for convenient use Related Product:LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant.
Especificaciones
Especificaciones
| Concentración | See Label |
| Para utilizar con (aplicación) | Kinase Assay |
| Formulación | 50mM sodium phosphate with 0.1mM PMSF, 300mM NaCl, 25% glycerol, 0.2mM DTT, 150mM Imidazole and no preservative; pH 7 |
| Peso molecular | 63.6-38-65 kDa |
| Nombre | Human AMPK alpha-1/beta-1/gamma-2, His Tag |
| Intervalo de pH | 7 |
| Método de purificación | Purified |
| Cantidad | 100 μg |
| Requisitos de almacenamiento | -80°C, Avoid Freeze/Thaw Cycles |
| Estado normativo | RUO |
| Mostrar más |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.