missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Gene ID (Entrez) | 55626 |
| Concentración | 1 mg/mL |
| Contenido y almacenamiento | Store at -80°C. Avoid freeze/thaw cycles. |
| Para utilizar con (aplicación) | This Peptide is Useful as a Blocking Peptide for NBP1-07124 |
| Alias de gen | activating molecule in BECN1-regulated autophagy protein 1, autophagy/beclin-1 regulator 1, DCAF3, DDB1 and CUL4 associated factor 3, FLJ20294, KIAA1736activating molecule in beclin-1-regulated autophagy, MGC33725, WD repeat domain 94, WDR94 |
| Símbolo de gen | AMBRA1 |
| Cantidad | 0,1 mg |
| Categoría de investigación | Autophagy |
| Tampón de almacenamiento | Peptide dissolved in dH20. Contains no BSA. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?