missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Synuclein Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Marca: Novus Biologicals NBP2-25146SS
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
alpha-Synuclein Polyclonal specifically detects alpha-Synuclein in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
Especificaciones
| alpha-Synuclein | |
| Polyclonal | |
| Unconjugated | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| Lewy body) 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor) | |
| Chicken | |
| 15 kDa | |
| 0.025 mL | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Neurodegeneration, Neuroscience | |
| 6622 | |
| Human, Mouse, Rat, Pig, Bovine, Equine | |
| IgY |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry Free-Floating 1:1000 | |
| P37840 | |
| SNCA | |
| This alpha-Synuclein Antibody was developed against Full length recombinant human alpha-Synuclein with the epitope from amino acids 61-95. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Rat and human. Human data from customer review. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido