missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 2-Macroglobulin Antibody (A2M/6554), DyLight 755, Novus Biologicals™
Tienda Bio Techne ProductosDescription
alpha 2-Macroglobulin Monoclonal antibody specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | alpha 2-Macroglobulin |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | A2M/6554 |
| Conjugado | DyLight 755 |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7 |
| Especie del huésped | Mouse |
| Inmunógeno | Human recombinant alpha 2-Macroglobulin protein fragment (around aa 604-748) (exact sequence is proprietary) |
| Método de purificación | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?