Learn More
ALKBH5 Antibody, Novus Biologicals™
Código de producto. p-200047703 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
25 μL
Tamaño de la unidad:
0.10 ml
25 microlitros
18464241 25 μL 25 microlitros
18708903 - 0.10 ml
2 options
Código de producto. 18464241
Marca: Novus Biologicals NBP18218825ul
Rabbit Polyclonal Antibody has been used in 6 publications
ALKBH5 Polyclonal specifically detects ALKBH5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno ALKBH5
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated
Formulación PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
N.º de referencia del gen Q6P6C2
Alias de gen ABH5OFOXD, alkB, alkylation repair homolog 5 (E. coli), Alkylated DNA repair protein alkB homolog 5, EC 1.14.11.-, FLJ20308, OFOXD1, oxoglutarate and iron-dependent oxygenase domain containing, probable alpha-ketoglutarate-dependent dioxygenase ABH5
Símbolos de los genes ALKBH5
Especie del huésped Rabbit
Inmunógeno This antibody was developed against Recombinant Protein corresponding to amino acids:EKLKSMTSRDNYKAGSREAAAAAAAAVAAAAAAAAAAEPYPVSGAKRKYQEDSDPERSDYEEQQLQKEEEARKVKSGIRQMRLFSQDECAKIEARIDEVVSRAEKGLYNEHTVD
Peso molecular del antígeno 51 kDa
Método de purificación Affinity Purified
Cantidad 25 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 54890
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only