missing translation for 'onlineSavingsMsg'
Learn More

Alkaline Phosphatase/ALPP Antibody (ALP/870) - IHC-Prediluted, Novus Biologicals™

Código de producto. 18129828 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
7 mL
Tamaño de la unidad:
7 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18129828 7 mL 7 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18129828 Proveedor Novus Biologicals N.º de proveedor NBP248092

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Alkaline Phosphatase/ALPP Monoclonal specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Alkaline Phosphatase/ALPP
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon ALP/870
Conjugado Unconjugated
Dilución Immunohistochemistry-Paraffin
Formulación 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Alias de gen Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme
Símbolos de los genes ALPP
Especie del huésped Mouse
Inmunógeno Recombinant full-length human Alkaline Phosphatase/ALPP protein (Uniprot: P05187 )
Peso molecular del antígeno 70 kDa
Método de purificación Protein A or G purified
Cantidad 7 mL
Estado normativo RUO
Disciplina de investigación Cancer, Embryonic Stem Cell Markers, Ovarian Carcinoma Cell Markers, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 250
Especificidad de la prueba Reacts with a 70kDa membrane-bound isozyme (Regan and Nagao type) of Placental Alkaline Phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. It is highly specific for PLAP and shows no cross-reaction with other isozymes of alkaline phosphatase. Anti-PLAP reacts with germ cell tumors and can discriminate between these and other neoplasms. Somatic neoplasms e.g. breast, gastrointestinal, prostatic, and urinary cancers may also immunoreact with antibodies to PLAP. Anti-PLAP positivity in conjunction with anti-keratin negativity favors seminoma over carcinoma. Germ cell tumors are usually anti-keratin positive, but they regularly fail to stain with anti-EMA, whereas most carcinomas stain with anti-EMA. Anti-PLAP has been useful in the diagnosis of gestational trophoblastic disease.
Especies diana Human
Contenido y almacenamiento Store at 4C.
Formulario Purified
Isotype IgG2b κ
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.