Learn More
Alix Antibody (3A9) - BSA Free, Novus Biologicals™
Código de producto. 18789923 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.10 miligramo
25 microgramos
18789923 0.025 mg 25 microgramos
18761134 0.1 mg 0.10 miligramo
2 options
Código de producto. 18789923
Marca: Novus Biologicals NB10065678SS
Mouse Monoclonal Antibody has been used in 4 publications
Alix Monoclonal specifically detects Alix in Human, Mouse, Rat, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Alix
Aplicaciones Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon 3A9
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulación Phosphate buffered saline with 0.09% Sodium Azide
N.º de referencia del gen Q8WUM4
Alias de gen AIP1DRIP4, ALG-2 interacting protein 1, ALG-2 interacting protein X, ALG-2-interacting protein 1, alinx, ALIX, apoptosis-linked gene 2-interacting protein X, dopamine receptor interacting protein 4, HP95, KIAA1375, MGC17003, PDCD6-interacting protein, programmed cell death 6 interacting protein, programmed cell death 6-interacting protein
Símbolos de los genes PDCD6IP
Especie del huésped Mouse
Inmunógeno Alix fusion protein
Método de purificación Protein G purified
Cantidad 0.025 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 10015
Especificidad de la prueba NB100-65678 recognizes apoptotic-linked-gene-product 2 (ALG-2) interacting protein X (ALIX), a conserved adaptor protein which is ubiquitously expressed. Alix was originally reported to play a role in apoptosis but has recently been shown to be involved in other cellular mechanisms including endosomal sorting, endocytosis, viral budding and actin cytoskeleton assembly.
Especies diana Human, Mouse, Rat, Xenopus species
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG1