Learn More
ALDOA Mouse anti-Human, Unconjugated, Clone: 1D6C2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67453-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The ALDOA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ALDOA. This antibody recognizes human, mouse, rat antigen. The ALDOA antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Especificaciones
| ALDOA | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 1D6C2 | |
| Unconjugated | |
| 150 μL | |
| 226 | |
| IgG1 |