Learn More
Tubo de Al-23, abierto en ambos extremos (paredes finas), D. E.: 12 mm, D. I.: 11 mm, Thermo Scientific™
44.80€ - 110.00€
Especificaciones
Nombre de nota | Both ends open (Thin walls) |
---|---|
Formulario | Tubo, ambos extremos abiertos (pared delgada), D.E.: 12 mm, D.I.: 11 mm |
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spezifikation
Both ends open (Thin walls) | |
Tubo, ambos extremos abiertos (pared delgada), D.E.: 12 mm, D.I.: 11 mm |
Sicherheit und Handhabung
missing translation for 'tsca' : U
Recommended Storage : Temperaturas ambientes