Learn More
AIF1 / Iba1 (Microglia Marker) Mouse, Clone: AIF1/1909, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 199-MSM1-P0
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| AIF1 / Iba1 (Microglia Marker) | |
| Cytoplasmic and Cell Surface | |
| AIF1/1909 | |
| Unconjugated | |
| Mouse | |
| 17kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 199 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Purified fragment of human recombinant AIF1 protein (apprx. aa 1-146) | |
| Lymph Node, Tonsil or Kidney | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |