Learn More
AIF-1/Iba1 Antibody (CL10930) [Biotin], Novus Biologicals™
Código de producto. 30296249 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30296249 0.1 mL 0.10 ml
1 options
Código de producto. 30296249 Proveedor Novus Biologicals N.º de proveedor NBP321184B
Mouse Monoclonal Antibody
AIF-1/Iba1 Monoclonal antibody specifically detects AIF-1/Iba1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno AIF-1/Iba1
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL10930
Conjugado Biotin
Formulación PBS
Alias de gen AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from P55008, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 199
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG2b