Learn More
AG-2/AGR2 Antibody (S04-1H6), Novus Biologicals™
Código de producto. 18698864 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18698864 50 μg 50 microlitros
18605864 100 μg 100 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18698864 Proveedor Novus Biologicals N.º de proveedor NBP31953450ul
Rabbit Monoclonal Antibody
AG-2/AGR2 Monoclonal antibody specifically detects AG-2/AGR2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno AG-2/AGR2
Aplicaciones Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon S04-1H6
Concentración 0.3 mg/mL
Conjugado Unconjugated
Dilución Western Blot, Immunohistochemistry 1:50, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulación 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Alias de gen AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog
Especie del huésped Rabbit
Inmunógeno A synthetic peptide of human AG-2/AGR2 (Uniprot # O95994)
Método de purificación Affinity purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Breast Cancer, Cancer, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 10551
Especies diana Human, Rat
Contenido y almacenamiento Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG