missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Adenovirus 3 Polyclonal antibody specifically detects Adenovirus 3 in Nonspecies specific, Adenovirus, Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | Adenovirus 3 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | PE |
| Dilución | Western Blot |
| Formulación | PBS |
| Alias de gen | ad3, adenovirus, Adenovirus 3 |
| Especie del huésped | Rabbit |
| Inmunógeno | Synthetic peptide within residues 250-270 of the fiber knob protein of Ad 3. |
| Método de purificación | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?