missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Adenosine A2aR Monoclonal antibody specifically detects Adenosine A2aR in Human, Mouse, Rat, Porcine, Canine, Guinea Pig, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Intracellular), CyTOF
Especificaciones
Especificaciones
| Antígeno | Adenosine A2aR |
| Aplicaciones | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | 7F6-G5-A2 |
| Concentración | 1.0 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:1000, Flow Cytometry 1-5 μg/mL, Immunohistochemistry 1 -5 μg/mL, Immunocytochemistry/ Immunofluorescence 1:50-1:100. Use reported in scientific literature (PMID 25836920), Immunohistochemistry-Paraffin 1 -5 μg/mL, Immunohistochemistry-Frozen reported in scientific literature (PMID 9822147), Flow (Intracellular) 1 - 5 μg/mL, CyTOF-ready |
| Formulación | PBS |
| N.º de referencia del gen | P29274, P30543, Q60613 |
| Alias de gen | adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?