missing translation for 'onlineSavingsMsg'
Learn More

Actin (Muscle Specific) Antibody (ACTA1/9105R), DyLight 550, Novus Biologicals™

Código de producto. 18599647 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18599647 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18599647 Proveedor Novus Biologicals N.º de proveedor NBP324312R

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Monoclonal Antibody

Actin (Muscle Specific) Monoclonal antibody specifically detects Actin (Muscle Specific) in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Actin (Muscle Specific)
Aplicaciones Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon ACTA1/9105R
Conjugado DyLight 550
Formulación 50 mM Sodium Borate
Alias de gen ACTA, ACTC, actin, alpha 2, smooth muscle, aorta, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, aortic smooth muscle, ACTSA, alpha-cardiac actin, ASD5, ASMA, Cell growth-inhibiting gene 46 protein, CFTD, CFTD1, CFTDM, CMD1R, CMH11, MPFD, NEM1, NEM2, NEM3, nemaline myopathy type 3
Especie del huésped Rabbit
Inmunógeno Recombinant human Actin (Muscle Specific) protein
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 58
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.