missing translation for 'onlineSavingsMsg'
Learn More

ACTH Antibody (CLIP/1407), Novus Biologicals™

Código de producto. 18151979 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.02 mg
0.1 mg
Tamaño de la unidad:
0.02 miligramo
0.10 miligramo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18151979 0.02 mg 0.02 miligramo
18107538 0.1 mg 0.10 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18151979 Proveedor Novus Biologicals N.º de proveedor NBP2476580.02MG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

ACTH Monoclonal specifically detects ACTH in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno ACTH
Aplicaciones Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence
Clasificación Monoclonal
Clon CLIP/1407
Conjugado Unconjugated
Dilución Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page, Immunofluorescence 1-2ug/ml
Formulación 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
N.º de referencia del gen P01189
Alias de gen ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein
Símbolos de los genes POMC
Especie del huésped Mouse
Inmunógeno Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF)
Método de purificación Protein A or G purified
Cantidad 0.02 mg
Estado normativo RUO
Disciplina de investigación Neuroscience, Nutrient Sensing in the Brain
Primario o secundario Primary
ID de gen (Entrez) 5443
Especificidad de la prueba ACTH (same as Corticotropin) is a 39 amino acid active peptide produced by the anterior pituitary. This MAb is specific to CLIP (aa25-39 of ACTH); does not react with Synacthen (aa1-24 of ACTH). POMC (pro-opiomelanocortin or corticotropin-lipotropin) is a 267 amino acid polypeptide hormone precursor that goes through extensive, tissue-specific posttranslational processing by convertases. POMC is cleaved into ten hormone chains named NPP, ACTH, alpha-MSH (Melanocyte Stimulating Hormone), beta-MSH, gamma-MSH, CLIP (corticotropin-like intermediary peptide), Lipotropin-beta, Lipotropin-gamma, beta-endorphin and Met-enkephalin. ACTH is also produced by cells of immune system (T-cells, B-cells, and macrophages) in response to stimuli associated with stress. Anti-ACTH is a useful marker in classification of pituitary tumors and the study of pituitary disease. It reacts with ACTH-producing cells (corticotrophs). It also may react with other tumors (e.g. some small cell carcinomas of the lung) causing paraneoplastic syndromes by secreting ACTH.
Especies diana Human
Contenido y almacenamiento Store at 4C.
Formulario Purified
Isotype IgG
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.