Descripción
- Used to purify antibodies from crude samples such as ascites fluid or immune serum
- Antibody to be purified or cleaned up ideally is in volume of 0.1mL to 0.5mL
- 20 to 500μg of antibody can be purified in each run
Especificaciones
Especificaciones
| Contenido y almacenamiento | 4°C |
| Cantidad | 1 Test |
| Contenido del kit | 4 or 12 additional collecting tubes, 1 vial AbPure wash buffer, 1 vial AbPure eution buffer, 1 vial AbPure neutralization buffer, 1 or 3 vials Protein A resin, 1 vial 10X AbPure binding buffer, 1/3 spin cartridge/collecting tube assemblies |
| Suficiente para | 1 purificación |
| Tipo de producto | Antibody Purification Kit |
Para uso exclusivo en investigación.