Learn More
Eurofins DiscoverX Abl (Q252H) Protein, Active
Descripción
The abl oncogene was initially identified as the viral transforming gene (v-abl) of Abelson murine leukemia virus (A-MuLV), from a chemically thymectomized mouse inoculated with Moloney murine leukemia virus (M-MuLV). The v-abl genome consists of M-MuLV gag sequences fused to mouse c-abl, resulting in the production of phosphoroteins, p160 and p120 gag/c-abl with protein-tyrosine kinase activity, as the major translational products. The abl oncogene is implicated in serval human leukemias including chronic myelocytic (CML), adult acute lymphoblastic (ALL) and pediatric ALL. In these leukemias the c-abl proto-oncogene undergoes a chromosomal translocation producing the Philadelphia (Ph¢) chromosome resulting in the fusion of c-abl sequences to the bcr gene of chromosome 22, and the molecular consequences of this translocation is the generation of a chimeric bcr/c-abl mRNA encoding activated abl protein-tyrosine kinase.
GenBank U07563
Especificaciones
Especificaciones
| Formulación | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 270 mM sucrose, 1 mMbenzamidine, 0.2 mM PMSF, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35. Frozen solution. |
| Peso molecular | 121 kDa |
| Cantidad | 10 μg |
| Especie | Human |
| Recombinante | Recombinant |
| Etiqueta de proteína | N-terminal 6His |
| Sistema de expresión | Recombinant enzyme expressed in Sf21 insect cells |
| Forma de proteína | Truncated |
| Grado de pureza o calidad | ≥42% |
| Expresión | Abl (Q252H), active |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.