missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ABCG2/CD338 Antibody (5D3) [Allophycocyanin/Cy7], Novus Biologicals™

Código de producto. 30027083 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
30027083 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30027083

Marca: Novus Biologicals™ FAB995APCCY7

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ABCG2/CD338 Monoclonal antibody specifically detects ABCG2/CD338 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ABCG2/CD338
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 5D3
Conjugado APC-Cyanine7
Formulación PBS
Alias de gen ABC transporter, ABC15, ABCPMGC102821, ATP-binding cassette sub-family G member 2, ATP-binding cassette transporter G2, ATP-binding cassette, sub-family G (WHITE), member 2, BCRP1, BCRPMRX, BMDP, Breast cancer resistance protein, CD338, CD338 antigen, CDw338, EST157481, mitoxantrone resistance protein, Mitoxantrone resistance-associated protein, MXRMXR1, placenta specific MDR protein, Placenta-specific ATP-binding cassette transporter
Especie del huésped Mouse
Inmunógeno 3T3 cells transduced with human ABCG2
Método de purificación Protein A or G purified from hybridoma culture supernatant
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación ABC Transporters, Cancer, Cellular Markers, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Stem Cell Markers, Stem Cells
Primario o secundario Primary
ID de gen (Entrez) 9429
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark. Do not freeze.
Formulario Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.