missing translation for 'onlineSavingsMsg'
Learn More

ABCB5 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Código de producto. 18583194 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18583194 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18583194 Proveedor Novus Biologicals N.º de proveedor NBP298737AF750

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABCB5 Polyclonal antibody specifically detects ABCB5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ABCB5
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Alexa Fluor 750
Formulación 50 mM Sodium Borate
Alias de gen ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human ABCB5.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación ABC Transporters, Lipid and Metabolism
Primario o secundario Primary
ID de gen (Entrez) 340273
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.