missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Benciloxibenzotrifluoruro, 95 %, Thermo Scientific™
17.20€ - 209.00€
Identificadores de productos químicos
CAS | 70097-65-3 |
---|---|
Fórmula molecular | C14H11F3O |
Peso molecular (g/mol) | 252.236 |
Número MDL | MFCD00275905 |
Clave InChI | SNRFAICCYVUTRG-UHFFFAOYSA-N |
Sinónimo | 1-benzyloxy-4-trifluoromethyl benzene, 4-benzyloxybenzotrifluoride, 4-benzyloxy benzotrifluoride, 1-phenylmethoxy-4-trifluoromethyl benzene, maybridge1_001022, 4 benzyloxy benzotrifluoride, 4-trifluoromethylphenyl benzyl ether, 1-benzyloxy-4-trifluoromethylbenzene, 1-benzyloxy-4-trifluoromethyl-benzene |
PubChem CID | 2798168 |
Nombre IUPAC | 1-fenilmetoxi-4-(trifluorometil)benceno |
SMILES | C1=CC=C(C=C1)COC2=CC=C(C=C2)C(F)(F)F |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
70097-65-3 | |
252.236 | |
SNRFAICCYVUTRG-UHFFFAOYSA-N | |
2798168 | |
C1=CC=C(C=C1)COC2=CC=C(C=C2)C(F)(F)F |
C14H11F3O | |
MFCD00275905 | |
1-benzyloxy-4-trifluoromethyl benzene, 4-benzyloxybenzotrifluoride, 4-benzyloxy benzotrifluoride, 1-phenylmethoxy-4-trifluoromethyl benzene, maybridge1_001022, 4 benzyloxy benzotrifluoride, 4-trifluoromethylphenyl benzyl ether, 1-benzyloxy-4-trifluoromethylbenzene, 1-benzyloxy-4-trifluoromethyl-benzene | |
1-fenilmetoxi-4-(trifluorometil)benceno |
Especificaciones
4-Benzyloxybenzotrifluoride | |
C14H11F3O | |
1-benzyloxy-4-trifluoromethyl benzene, 4-benzyloxybenzotrifluoride, 4-benzyloxy benzotrifluoride, 1-phenylmethoxy-4-trifluoromethyl benzene, maybridge1_001022, 4 benzyloxy benzotrifluoride, 4-trifluoromethylphenyl benzyl ether, 1-benzyloxy-4-trifluoromethylbenzene, 1-benzyloxy-4-trifluoromethyl-benzene | |
C1=CC=C(C=C1)COC2=CC=C(C=C2)C(F)(F)F | |
252.236 | |
252.23 | |
Sólido |
70097-65-3 | |
MFCD00275905 | |
SNRFAICCYVUTRG-UHFFFAOYSA-N | |
1-fenilmetoxi-4-(trifluorometil)benceno | |
2798168 | |
95% |
Seguridad y manipulación
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes