missing translation for 'onlineSavingsMsg'
Learn More

4-1BB/TNFRSF9/CD137 Antibody (2356B), mFluor Violet 450 SE, Novus Biologicals™

Código de producto. 18590499 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
18590499 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18590499

Marca: Novus Biologicals FAB8381MFV450

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Monoclonal Antibody

4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno 4-1BB/TNFRSF9/CD137
Aplicaciones Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Clasificación Monoclonal
Clon 2356B
Conjugado mFluor Violet 450 SE
Dilución Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulación 50mM Sodium Borate
Alias de gen 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Especie del huésped Rabbit
Inmunógeno Chinese hamster ovary cell line CHO-derived recombinant human 4-1BB/TNFRSF9/CD137, Leu24-His183, Accession # Q07011
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells
Primario o secundario Primary
ID de gen (Entrez) 3604
Especies diana Human
Contenido y almacenamiento Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.