Learn More
3-cloro-4-(trifluorometoxi)anilina, 97 %, Thermo Scientific™
53.00€ - 189.00€
CAS | 64628-73-5 |
---|---|
Fórmula molecular | C7H5ClF3NO |
Peso molecular (g/mol) | 211.568 |
Número MDL | MFCD00190126 |
Clave InChI | ZPKUUNGPBSRPRM-UHFFFAOYSA-N |
Sinónimo | 3-chloro-4-trifluoromethoxy aniline, benzenamine, 3-chloro-4-trifluoromethoxy, 3-chloro-4-trifluoromethoxy phenylamine, pubchem2949, acmc-1b4hb, ksc496e4r, 3-chloro4-trifluoromethoxyaniline, 3-chloro-4-trifluoromethoxy-aniline, 3-chloro-4-trifluoromethoxy-phenylamine |
PubChem CID | 2773843 |
Nombre IUPAC | 3-cloro-4-(trifluorometoxi)anilina |
SMILES | C1=CC(=C(C=C1N)Cl)OC(F)(F)F |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
11475667
|
Thermo Scientific Alfa Aesar
B23630.04 |
2 g |
53.00€
2 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
11485667
|
Thermo Scientific Alfa Aesar
B23630.09 |
10 g | |||||||
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications3-Chloro-4-(trifluoromethoxy)aniline is used as a primary and secondary intermediate in chemical synthesis.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
64628-73-5 | |
211.568 | |
ZPKUUNGPBSRPRM-UHFFFAOYSA-N | |
2773843 | |
C1=CC(=C(C=C1N)Cl)OC(F)(F)F |
C7H5ClF3NO | |
MFCD00190126 | |
3-chloro-4-trifluoromethoxy aniline, benzenamine, 3-chloro-4-trifluoromethoxy, 3-chloro-4-trifluoromethoxy phenylamine, pubchem2949, acmc-1b4hb, ksc496e4r, 3-chloro4-trifluoromethoxyaniline, 3-chloro-4-trifluoromethoxy-aniline, 3-chloro-4-trifluoromethoxy-phenylamine | |
3-cloro-4-(trifluorometoxi)anilina |
Especificaciones
3-Chloro-4-(trifluoromethoxy)aniline | |
64628-73-5 | |
C7H5ClF3NO | |
6314199 | |
Slightly soluble in water. | |
C1=CC(=C(C=C1N)Cl)OC(F)(F)F | |
211.568 | |
211.57 |
29°C to 30°C | |
1.454 | |
MFCD00190126 | |
3-chloro-4-trifluoromethoxy aniline, benzenamine, 3-chloro-4-trifluoromethoxy, 3-chloro-4-trifluoromethoxy phenylamine, pubchem2949, acmc-1b4hb, ksc496e4r, 3-chloro4-trifluoromethoxyaniline, 3-chloro-4-trifluoromethoxy-aniline, 3-chloro-4-trifluoromethoxy-phenylamine | |
ZPKUUNGPBSRPRM-UHFFFAOYSA-N | |
3-cloro-4-(trifluorometoxi)anilina | |
2773843 | |
97% |
Seguridad y manipulación
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes