missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Bromofenil)-4-(3,4-dimetoxifenil)-5-metil-4H-1,2,4-triazol, 97 %, Thermo Scientific™
Identificadores de productos químicos
CAS | 1858255-54-5 |
---|---|
Fórmula molecular | C17H16BrN3O2 |
Peso molecular (g/mol) | 374.238 |
Número MDL | MFCD22683127 |
Clave InChI | SPFFEOOTWDLJSH-UHFFFAOYSA-N |
Sinónimo | 3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-4h-1,2,4-triazole |
PubChem CID | 98000573 |
Nombre IUPAC | 3-(4-bromofenil)-4-(3,4-dimetoxifenil)-5-metil-1,2,4-triazol |
SMILES | CC1=NN=C(N1C2=CC(=C(C=C2)OC)OC)C3=CC=C(C=C3)Br |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
1858255-54-5 | |
374.238 | |
SPFFEOOTWDLJSH-UHFFFAOYSA-N | |
98000573 | |
CC1=NN=C(N1C2=CC(=C(C=C2)OC)OC)C3=CC=C(C=C3)Br |
C17H16BrN3O2 | |
MFCD22683127 | |
3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-4h-1,2,4-triazole | |
3-(4-bromofenil)-4-(3,4-dimetoxifenil)-5-metil-1,2,4-triazol |
Especificaciones
3-(4-Bromophenyl)-4-(3,4-dimethoxyphenyl)-5-methyl-4H-1,2,4-triazole | |
C17H16BrN3O2 | |
3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-1,2,4-triazole, 3-4-bromophenyl-4-3,4-dimethoxyphenyl-5-methyl-4h-1,2,4-triazole | |
CC1=NN=C(N1C2=CC(=C(C=C2)OC)OC)C3=CC=C(C=C3)Br | |
374.238 | |
374.24 |
1858255-54-5 | |
MFCD22683127 | |
SPFFEOOTWDLJSH-UHFFFAOYSA-N | |
3-(4-bromofenil)-4-(3,4-dimetoxifenil)-5-metil-1,2,4-triazol | |
98000573 | |
97% |
Seguridad y manipulación
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes