missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Clorhidrato de deoxicitidina, 98 %, Thermo Scientific Chemicals
Identificadores de productos químicos
CAS | 3992-42-5 |
---|---|
Fórmula molecular | C9H14ClN3O4 |
Peso molecular (g/mol) | 263.68 |
Número MDL | MFCD00012840 |
Clave InChI | LTKCXZGFJFAPLY-VOBHXVAMNA-N |
Sinónimo | 2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl |
PubChem CID | 107488 |
Nombre IUPAC | 4-amino-1-[(2R,4S,5R)-4-hidroxi-5-(hidroximetil)oxolan-2-il]pirimidin-2-ona; clorhidrato |
SMILES | Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1 |
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
3992-42-5 | |
263.68 | |
LTKCXZGFJFAPLY-VOBHXVAMNA-N | |
107488 | |
Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1 |
C9H14ClN3O4 | |
MFCD00012840 | |
2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl | |
4-amino-1-[(2R,4S,5R)-4-hidroxi-5-(hidroximetil)oxolan-2-il]pirimidin-2-ona; clorhidrato |
Especificaciones
2'-Deoxycytidine hydrochloride | |
Auténtico | |
Frasco de vidrio | |
MFCD00012840 | |
LTKCXZGFJFAPLY-VOBHXVAMNA-N | |
4-amino-1-[(2R,4S,5R)-4-hidroxi-5-(hidroximetil)oxolan-2-il]pirimidin-2-ona; clorhidrato | |
107488 | |
98% |
3992-42-5 | |
97.5% min. (HPLC) | |
C9H14ClN3O4 | |
2'-deoxycytidine hydrochloride, deoxycytidine hydrochloride, cytidine, 2'-deoxy-, monohydrochloride, 2'-deoxycytidine hcl, unii-x8fx60e66d, 2'-deoxycytidine monohydrochloride, cytidine, 2'-deoxy-, hydrochloride 1:1, cytosine deoxyriboside hydrochloride, dc.hcl | |
Cl.NC1=NC(=O)N(C=C1)[C@H]1C[C@H](O)[C@@H](CO)O1 | |
263.68 | |
263.68 |
Seguridad y manipulación
missing translation for 'einecsNumber' : 223-639-9
RUO – Research Use Only