missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, ACS Reagent, ≥99%, Honeywell Riedel-de Haën™
Click to view available options
Cantidad:
500 mL
1 L
2 L
2.5 L
20 L
6 x 1 L
Envase:
Frasco de vidrio
Glass Bottle
Stainless Steel Can
Descripción
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Especificaciones
Punto de fusión | -87°C |
Densidad | 0.805g/mL at 25°C |
Punto de ebullición | 80°C |
Cantidad | 500 mL |
Temperatura de inflamación | −3°C (26.6°F) |
Espectro infrarrojo | Conforms to structure |
Envase | Glass Bottle |
Índice de refracción | n20/D 1.379 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'