missing translation for 'onlineSavingsMsg'
Learn More

Ferritin Mouse anti-Human, Clone: FTH/2081, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP3-07795-100ug

 Ver más versiones de este producto

Código de producto. 18659248

  • 521.00€ / 100 microgramos

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Description

Description

Ferritin Monoclonal specifically detects Ferritin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ferritin
Monoclonal
0.2 mg/mL
Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 0.1-0.2 μg/mL
apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15
Recombinant human Ferritin protein fragment (around aa 58-180) (exact sequence is proprietary)
100 μg
Cancer, Cellular Markers, Lipid and Metabolism
2495
Store at 4°C.
IgG2c κ
Western Blot, Immunohistochemistry (Paraffin)
FTH/2081
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.