missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Boc-1-piperidinil)-4-bromopirazol, 98 %, Thermo Scientific Chemicals
26.22€ - 334.00€
Identificadores de productos químicos
CAS | 877399-50-3 |
---|---|
Fórmula molecular | C13H20BrN3O2 |
Peso molecular (g/mol) | 330.23 |
Número MDL | MFCD09831980 |
Clave InChI | IYNZAVDBHAQODX-UHFFFAOYSA-N |
Sinónimo | tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-bromopyrazol-1-yl piperidine, 4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester, 4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate, n-boc-4-4-bromopyrazol-1-yl piperidine, 3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, ksc658s5d, 1-boc-4-4-bromo-1-pyrazolyl piperidine |
PubChem CID | 45480278 |
Nombre IUPAC | 4-(4-bromopirazol-1-il)piperidina-1-carboxilato de terc-butilo |
SMILES | CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1 |
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
877399-50-3 | |
330.23 | |
IYNZAVDBHAQODX-UHFFFAOYSA-N | |
45480278 | |
CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1 |
C13H20BrN3O2 | |
MFCD09831980 | |
tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-bromopyrazol-1-yl piperidine, 4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester, 4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate, n-boc-4-4-bromopyrazol-1-yl piperidine, 3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, ksc658s5d, 1-boc-4-4-bromo-1-pyrazolyl piperidine | |
4-(4-bromopirazol-1-il)piperidina-1-carboxilato de terc-butilo |
Especificaciones
1-(4-Boc-1-piperidinyl)-4-bromopyrazole | |
Blanco | |
MFCD09831980 | |
tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-bromopyrazol-1-yl piperidine, 4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester, 4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate, n-boc-4-4-bromopyrazol-1-yl piperidine, 3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, ksc658s5d, 1-boc-4-4-bromo-1-pyrazolyl piperidine | |
CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1 | |
330.23 | |
330.23 | |
Sólido |
877399-50-3 | |
C13H20BrN3O2 | |
14511017 | |
IYNZAVDBHAQODX-UHFFFAOYSA-N | |
4-(4-bromopirazol-1-il)piperidina-1-carboxilato de terc-butilo | |
45480278 | |
98% |
Seguridad y manipulación
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes
RUO – Research Use Only