missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
HLA ABC Monoclonal antibody specifically detects HLA ABC in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antígeno | HLA ABC |
| Aplicaciones | Flow Cytometry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | Bra-23/9 |
| Conjugado | DyLight 350 |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 |
| Especie del huésped | Mouse |
| Inmunógeno | Human REH cells (pre-B leukaemia cell line) |
| Método de purificación | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?