missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NB100-56176
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TRAF-3 Polyclonal specifically detects TRAF-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Especificaciones
| TRAF-3 | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:4000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:4000 | |
| CAP-1CD40 receptor associated factor 1, CD40 associated protein 1, CD40 binding protein, CD40 receptor-associated factor 1, CD40-binding protein, CD40bp, CRAF1CAP1, LAP1CD40BP, LMP1-associated protein 1, TNF receptor-associated factor 3 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| TRAF3 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.02% Sodium Azide | |
| TRAF3 | |
| A recombinant protein fragment corresponding to the cytoplasmic end of human TRAF3 (amino acids 341-568) was used as immunogen, GenBank no. GenBank no. gi 22027616 ref NP_003291.2 . | |
| 0.05 mL | |
| Apoptosis, Immunology, Innate Immunity, Zinc Finger | |
| 7187 | |
| Human, Mouse, Rat | |
| Antisera |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido