CAS RN 107650-06-6
CAS RN 107650-06-6
IUPAC Name:
4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluorononane-1,2-diol
Sinónimos:
4,4,5,5,6,6,7,7,8,8,9,9,9-tridecafluorononane-1,2-diol
Peso molecular (g/mol):
394.13
Fórmula molecular:
C9H7F13O2
InChi Key:
DAHZCNRVTNHGGR-UHFFFAOYNA-N
SMILES:
OCC(O)CC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F