CAS RN 53215-95-5
CAS RN 53215-95-5
IUPAC Nombre:
benzyl[(trimethylsilyl)methyl]amine
Sinónimos:
benzyl[(trimethylsilyl)methyl]amine
Peso molecular (g/mol):
193.37
Fórmula molecular:
C11H19NSi
InChi Key:
WECLUYCAWLJMKM-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)CNCC1=CC=CC=C1
1
–
1
de
1
resultados
1
N-[(trimetilsilil)metil]benzilamina, 95 %, Thermo Scientific Chemicals
CAS: 53215-95-5 Fórmula molecular: C11H19NSi Peso molecular (g/mol): 193.37 Número MDL: MFCD00012192 Clave InChI: WECLUYCAWLJMKM-UHFFFAOYSA-N Sinónimo: n-trimethylsilylmethyl benzylamine,n-trimethylsilyl methyl benzylamine,benzyl trimethylsilyl methyl amine,n-benzyl-1-trimethylsilyl methylamine,1-phenyl-n-trimethylsilylmethyl methanamine,benzyl trimethylsilylmethyl amine,n-benzyl-1-trimethylsilyl methanamine,benzenemethanamine, n-trimethylsilyl methyl,n-benzylaminomethyltrimethylsilane,n-benzyl-n-trimethylsilyl methyl amine PubChem CID: 353458 SMILES: C[Si](C)(C)CNCC1=CC=CC=C1