Learn More
Invitrogen™ ZONAB Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ 402800
Descripción
This antibody reacts with an internal region of the ZONAB protein. On Western blots, it identifies a major band at ~65 kDa and a minor band at ~54 kDa. The antibody is directed towards a region of ZONAB common to both the long and the alternatively-spliced short form. During development, reactivity was confirmed using mouse testis tissue homogenates by Western blotting and labeling of mouse testis tissue using immunofluorescence. All signals were absent in knockout mice, further confirming the reactivity with ZONAB.
Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.
Especificaciones
| ZONAB | |
| Polyclonal | |
| Unconjugated | |
| YBX3 | |
| cold shock domain protein A; cold shock domain-containing protein A; cold-shock domain containing A1; cold-shock domain protein A; Csda; CSDA1; Dbpa; DNA-binding protein A; Dpba; MSY3; MSY4; Muscle Y-box protein YB2; oxyR; RYB-A; single-strand DNA-binding protein NF-GMB; Y box binding protein 3; Y box protein 3; Yb2; Y-box binding protein 3; Y-box protein 3; Y-box-binding protein 3; Y-box-binding protein A; Ybx3; ZO-1-associated nucleic acid-binding protein; ZONAB; ZONAB A; ZONAB B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56449, 83807 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Q62764, Q9JKB3 | |
| YBX3 | |
| Synthetic peptide derived from an internal region of mouse ZONAB (ZO-1 associated nucleic acid-binding protein. | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.