Learn More
Invitrogen™ ZO-1 Monoclonal Antibody (ZO1-1A12), Alexa Fluor™ 488
Descripción
ZO-1 Monoclonal Antibody for ICC/IF
ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.
Especificaciones
Especificaciones
| Antígeno | ZO-1 |
| Aplicaciones | Immunocytochemistry |
| Clasificación | Monoclonal |
| Clon | ZO1-1A12 |
| Concentración | 1 mg/mL |
| Conjugado | Alexa Fluor 488 |
| Formulación | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| génica | TJP1 |
| N.º de referencia del gen | O97758, Q07157 |
| Alias de gen | DKFZp686M05161; I79_005485; MGC133289; OTTHUMP00000174520; Tight junction protein 1; tight junction protein 1 (zona occludens 1); tight junction protein ZO-1; Tjp1; zo1; ZO-1; ZO-1 MDCK; zona occludens 1; zona occludens protein 1; zonula occludens 1; zonula occludens 1 protein; zonula occludens protein 1 |
| Mostrar más |