Learn More
ZNF81 Antibody (PCRP-ZNF81-2C7) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals (Bio-Techne) NBP3-14081JF525
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ZNF81 Monoclonal antibody specifically detects ZNF81 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Western Blot,Protein Array,Immunocytochemistry/ Immunofluorescence
Especificaciones
| ZNF81 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| dJ54B20.6, FLJ44367, HFZ20X-linked 45, zinc finger protein 81, zinc finger protein 81 (HFZ20) | |
| Recombinant full-length human ZNF81 protein (Uniprot: P51508) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Western Blot, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-ZNF81-2C7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 347344 | |
| Store at 4°C in the dark. | |
| IgG2b |