missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF460 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25299-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF460 (zinc finger protein 460), also known as HZF8 or ZNF272, is a 562 amino acid nuclear protein that is ubiquitously expressed at low levels and expressed at high levels in pancreas and liver. Belonging to the Krueppel C2 H2-type zinc-finger protein family, ZNF460 contains 11 C2 H2-type zinc fingers and a KRAB domain. Like other zinc-fingers, ZNF460 is also suggested to be involved in transcriptional regulation.Especificaciones
| ZNF460 | |
| Polyclonal | |
| Unconjugated | |
| ZNF460 | |
| HZF8, Zinc finger protein 272, zinc finger protein 460, Zinc finger protein HZF8, ZNF272, ZNF460 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10794 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14592 | |
| ZNF460 | |
| ZNF460 Fusion Protein Ag18106 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido