missing translation for 'onlineSavingsMsg'
Learn More

ZNF451 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

140.00€ - 399.00€

Especificaciones

Antígeno ZNF451
Concentración 0.31 mg/mL
Aplicaciones Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
Clasificación Polyclonal
Conjugado Unconjugated
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16803805
Ver documentos Detalles de la promoción
Proteintech
25228-1-AP-20UL
20 μL
140.00€
20 microlitros
Ver productos alternativos
Ahorre hasta 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16893795
Ver documentos Detalles de la promoción
Proteintech
25228-1-AP-150UL
150 μL
399.00€
150 microlitros
Ver productos alternativos
Ahorre hasta 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

RNA polymerase (pol) III synthesizes tRNA, 5s rRNA, 7SL RNA and U6 snRNA and is overexpressed in many transformed cell lines and tumors in vivo, since cells must duplicate its protein components before division. Therefore, in order to maintain rapid growth, cells must produce a high level of Pol III transcribed RNA, which requires the presence of the TFIIIB and TFIIIC2 transcription factor complexes. The TFIIIC2 complex is composed of five subunits, TFIIIC220, TFIIIC110, TFIIIC102, TFIIIC90 and TFIIIC63, that are overexpressed in adenovirus transformed cells as well as in maligt cells in vivo, such as ovarian carcinomas. TFIIIC2 recruits RNA pol III and TFIIIB to promoter elements and may be a key component in the deregulation of maligt cells. The TFIIIB complex includes the TATA-binding protein (TBP), TFIIB-related factor 1 (BRF1) and TFIIIB, the expression of which are also upregulated in transformed cells. In many carcinomas, the tumor suppressors retinoblastoma (RB) and p53 are inactivated, which affects their ability to bind and inactivate the function of TFIIIB.
Especificaciones

Especificaciones

ZNF451
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
Rabbit
Human
Q9Y4E5
26036
ZNF451 Fusion Protein Ag18346
Primary
-20°C
ZNF451
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
COASTER, KIAA0576, KIAA1702, zinc finger protein 451, ZNF451
ZNF451
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documentos

Documentos

Product Certifications