missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF418 Polyclonal Antibody

Description
Peptide sequence: HRCEAWGNKL YDSSNRPHQN QYLGEKPYRS SVEEALFVKR CKFHVSEESS Sequence homology: Human: 100%.
ZNF418 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 16 C2H2-type zinc fingers and 1 KRAB domain. ZNF418 is a transcriptional repressor.
Specifications
Specifications
| Antígeno | ZNF418 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.5 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 2% sucrose and 0.09% sodium azide |
| génica | ZNF418 |
| N.º de referencia del gen | Q8TF45 |
| Alias de gen | FLJ31551; KIAA1956; MGC138449; Zinc finger protein 418; ZNF418 |
| Símbolos de los genes | ZNF418 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?