missing translation for 'onlineSavingsMsg'
Learn More

ZNF2 Monoclonal Antibody (OTI2A9), TrueMAB™, OriGene

Código de producto. 17574614
Encompass_Preferred
Change view
Click to view available options
Cantidad:
100 μL
30 μL
Tamaño de la unidad:
100 microlitros
30 microlitros
Código de producto. Cantidad unitSize
17574614 30 μL 30 microlitros
17554614 100 μL 100 microlitros
2 options
This item is not returnable. View return policy

Product Code. 17574614

Brand: Origene Technologies TA811017S

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF2 Monoclonal antibody specifically detects ZNF2 in Human samples. It is validated for Western Blot

Specifications

Antígeno ZNF2
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI2A9
Concentración 1 mg/mL
Conjugado Unconjugated
Formulación PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
génica ZNF2
N.º de referencia del gen Q9BSG1
Alias de gen 2810405K07Rik; A1-5; Mkr2; Mkr-2; multifinger protein mKr2; Zfp2; Zfp661; zfp-661; zinc finger protein 2; zinc finger protein 2 (A1-5); zinc finger protein 2 homolog; zinc finger protein 2.2; Zinc finger protein 661; ZNF2; ZNF661
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human ZNF2 produced in E.coli.
Método de purificación Affinity chromatography
Cantidad 30 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7549
Especies diana Human
Contenido y almacenamiento -20° C, Avoid Freeze/Thaw Cycles
Formulario Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.