missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA541410
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide sequence: ADECSGCGKS LLHIKLEKTH PGDQAYEFNQ NGEPYTLNEE SLYQKIRILE Sequence homology: Dog: 86%; Horse: 79%; Human: 100%; Rat: 100%.
Two members of the human zinc finger Kruppel family, ZNF 12 (KOX 3) and ZNF 26 (KOX 20), have been localized by somatic cell hybrid analysis and in situ chromosomal hybridization. The presence of individual human zinc finger genes in mouse-human hybrid DNAs was correlated with the presence of specific human chromosomes or regions of chromosomes in the corresponding cell hybrids. Analysis of such mouse-human hybrid DNAs assigned the ZNF 12 (KOX 3) gene to chromosome region 7p.
Especificaciones
| ZNF12 | |
| Polyclonal | |
| Unconjugated | |
| Znf12 | |
| C530015C18; GIOT3; GIOT-3; gonadotropin inducible transcription repressor 3; gonadotropin-inducible ovary transcription repressor 3; HZF11; KOX3; Krox-7; Zfp12; Zfp-12; zinc finger protein 11; zinc finger protein 12; Zinc finger protein 325; zinc finger protein KOX3; Znf12; ZNF325 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7559 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P17014 | |
| Znf12 | |
| Synthetic peptide directed towards the N-terminal of human ZNF12 (aa 165-214). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido