Learn More
Chemsplash Zip Labcoat T5/6 Fabric, Elastic Cuff
Descripción
Chemsplash Lab Coat
- Chemsplash 55GSM Type PB 6B Fabric
- Zip Fasten Front with Adhesive Flap Cover
- Elasticated Cuffs
- Fold Over Collar
- Silicone and Latex Free
- Anti-Static Material
- Non-Linting
- PFAS Free
- Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
Description: The Chemsplash Lab Coat is made from Chemsplash fabric, which is soft and microporous. It is strong and resistant to chemical splash and dust. This lab coat is a 2/3 length coat with a collar, zip closure, and elasticated wrists. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from the environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Especificaciones
Especificaciones
| Tipo de producto | Lab Coat |
| Tamaño | X-Large |
| Sexo | Unisex |
| Material | Microporous Composite Fabric |
| Color | Blanco |
| Tipo de tapón | Zipper |
| Estilo de manguito | Elástico |
| Desechables | Single-use |
| Esterilidad | No estéril |
| Resistencia | Liquids |
| Mostrar más |